Learn More
Electron Microscopy Sciences Anti Static Brush, Short
SDP
Catalog No. 5024559
Encompass_Preferred
Eliminates static electric charges and removes dust, lint and other particles from negatives and lenses with one quick operation. These non-chemical, non-radioactive, synthetic brushes use the natural principal of resistance flow to obliterate dust and the static electricity that attracts it.
They are absolutely safe on all photographic surfaces and emulsions, great for computer monitors and glass screens as well. They can be cleaned with soap and rinsed with water. They never need "recharging" and will last a lifetime with proper care.
Catalog No. 50-245-59 Supplier Electron Microscopy Sciences Supplier No. 6061204
Eliminates static electric charges and removes dust, lint and other particles from negatives and lenses with one quick operation. These non-chemical, non-radioactive, synthetic brushes use the natural principal of resistance flow to obliterate dust and the static electricity that attracts it.
They are absolutely safe on all photographic surfaces and emulsions, great for computer monitors and glass screens as well. They can be cleaned with soap and rinsed with water. They never need "recharging" and will last a lifetime with proper care.