Learn More
Mopec Anti-static Bubble Bag
SDP
Catalog No. 22446140 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
6 x 8 in.
8 x 11.5 in.
12 x 15.5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-140 12 x 15.5 in.
22-446-108 6 x 8 in.
22-446-100 8 x 11.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-140 Supplier Mopec Supplier No. FS080
Add to Cart
Add to Cart
Ideal for secure handling and storage, each bag features a resealable closure with the option for heat sealing.
  • Self-sealing glue strip
  • Superior cushioning for delicate items
  • Keeps electronic devices safe from static
  • Can be Heat-sealed with an impulse sealer
  • Anti-static bubble bags designed to protect sensitive electronic devices from static damage while providing reliable cushioning.
Specifications
Dimensions (L x W) 12 x 15.5 in.
Product Type Evidence Bag
Color Pink
Quantity 100/Pk.