Learn More
MSC Bosch Dust Extractor Hose, Anti-static, 16.4 ft, 35 mm Diameter
Catalog No. 502839288
Encompass
This anti-static dust extractor hose is designed for use with compatible dust extractors. It features a crush-resistant and flexible design, and includes a locking ring for secure connection to power tool adapters and dust collection guards/shrouds.
  • Anti-static, crush resistant replacement hose
  • Locking ring securely connects to power tool adapter and dust collection guards/shrouds
  • Flexible hose, 16.4 ft in length
  • Includes anti-static adapter for connection to power tools
Catalog No. 50-283-9288 Supplier MSC Supplier No. 58149857
This anti-static dust extractor hose is designed for use with compatible dust extractors. It features a crush-resistant and flexible design, and includes a locking ring for secure connection to power tool adapters and dust collection guards/shrouds.
  • Anti-static, crush resistant replacement hose
  • Locking ring securely connects to power tool adapter and dust collection guards/shrouds
  • Flexible hose, 16.4 ft in length
  • Includes anti-static adapter for connection to power tools