Learn More
Perkin Elmer US LLC Anti-Static Glass Compound - Qty. 8 oz.
Catalog No. 502016472
Encompass_Preferred
Anti-Static Glass Compound comes in an 8 oz tube. It can be applied to the outside of glassware to reduce static electricity. Qty. 8 oz
Catalog No. 50-201-6472 Supplier Perkin Elmer US LLC Supplier No. 09905932
Anti-Static Glass Compound comes in an 8 oz tube. It can be applied to the outside of glassware to reduce static electricity. Qty. 8 oz