Learn More
Bellex International Corporation Zerostat 3 anti-static gun
SDP
Catalog No. NC9663078
Encompass_Preferred
•Effective in neutralizing electrostatic charges on virtually any surface. •Eliminates static electricity in the laboratory, plant, darkroom or home. ◾Prevents charged particles from flying during weighing or transfer of powders and keeps powders from sticking to containers. ◾Keeps film, glass, laboratory slides and plasticware dust free. ◾Neutralizes static from photographic lenses, negatives and slides to release dust and lint. ◾Constructed from durable hard plastic, a long lasting alternative to cleaning brushes. ◾No batteries or electrical power supply required. ◾Non-radioactive. ◾Convenient, hand-held and lightweight.
Catalog No. NC9663078 Supplier Bellex International Corporation Supplier No. ZEROSTAT3
•Effective in neutralizing electrostatic charges on virtually any surface. •Eliminates static electricity in the laboratory, plant, darkroom or home. ◾Prevents charged particles from flying during weighing or transfer of powders and keeps powders from sticking to containers. ◾Keeps film, glass, laboratory slides and plasticware dust free. ◾Neutralizes static from photographic lenses, negatives and slides to release dust and lint. ◾Constructed from durable hard plastic, a long lasting alternative to cleaning brushes. ◾No batteries or electrical power supply required. ◾Non-radioactive. ◾Convenient, hand-held and lightweight.