Learn More
Uline Anti-Static Table Mat, Vinyl
Supplier:  Uline S12743
Encompass
Protect your electronic devices Transfers static charges from work surface to ground. Mats include two snap fasteners and grounding cord for wrist straps. 3-layer construction General packaging and assembly use. Economy and Deluxe Grounding Wrist Straps sold separately. MEETS ESD STANDARD 41 ESD STANDARD 2020 EIA625 MIL-STD-1686. Grounding cord included. Two grounding snaps come installed. Mat can be cut without affecting anti-static properties. Use an ESD mat cleaner that does not contain silicone.
Catalog No. NC1580479