Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Anti-Static Nitrile Finger Cots

Catalog No. 191502137
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-2137 Small
19-150-2138 Medium
19-150-2139 Large
19-150-2140 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-150-2137 Supplier Valutek™ Supplier No. VTCNRPFSM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disposable, powder-free finger cots are inherently static dissipative, and constructed of 100% clean synthetic nitrile polymer to eliminate allergy risks caused by natural rubber latex.

  • Constructed from 100% clean, synthetic nitrile polymer
  • Free of allergy-causing rubber latex, fillers, and silicones
  • Finger cots are inherently static dissipative and suitable for ESD-sensitive components
  • Recommended for use in a cleanroom Class 100-1000 (ISO 5-6) critical environment
  • Ideal for handling Class II ESD sensitive devices with thresholds above 100 V
  • Tested and manufactured in ISO-compliant facilities under strict processes to ensure product specifications

Specifications

Cleanroom Class Class 100-1000 (ISO 5-6)
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Acrylonitrile Butadiene
Powdered Powder Free
Product Type ESD Finger Cot
Size Small
For Use With (Application) Cleanroom Class 100-1000 (ISO 5-6) Critical Environments
For Use With (Equipment) Class II ESD Sensitive Devices
Glove Cuff Style Beaded
Manufacturing/QMS Audit Standards Manufactured in ISO-compliant facilities
Packaging Type Bag
Series VTCNRPF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.