Learn More
Uline 12 x 18" 4 Mil Clear Anti-Static Poly Bags
Supplier:  Uline S13021
Encompass_Preferred
12 x 18" 4 Mil Clear Anti-Static Poly Bags 500 per carton. Same static protection as pink poly bags with clear view of product. Noncorrosive virgin polyethylene film. Heat sealable. Not tested for weight capacity.
Catalog No. NC3137362