Learn More
BURKLE INC Anti-static set in case (3 copper cable with clamps)
Catalog No. NC3451724
Encompass_Preferred
When decanting flammable liquids without the machines and containers involved in the process being sufficiently earthed, electrostatic build-up can occur, which for many substances can result in fire or an explosion.
Bürkle’s Anti-static set ensures easy and reliable earthing.
The Anti-static set consists of three colour-coded connecting copper cables with clamps in a handy plastic case.
Catalog No. NC3451724 Supplier BURKLE INC Supplier No. 56021000
When decanting flammable liquids without the machines and containers involved in the process being sufficiently earthed, electrostatic build-up can occur, which for many substances can result in fire or an explosion.
Bürkle’s Anti-static set ensures easy and reliable earthing.
The Anti-static set consists of three colour-coded connecting copper cables with clamps in a handy plastic case.