Learn More
Uline Anti-Static Table Mat, Vinyl
Catalog No. NC1460407
Encompass
Grounding cord included, Two grounding snaps come installed, Mat can be cut without affecting anti-static properties
Catalog No. NC1460407 Supplier Uline Supplier No. S12744
Grounding cord included, Two grounding snaps come installed, Mat can be cut without affecting anti-static properties