Learn More
Cole-Parmer Anti-Static Weighing Funnel, PP, Medium; 50/PK
Catalog No. NC9026536 Shop All Cole-Parmer Products
Encompass_Preferred
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.
Catalog No. NC9026536 Supplier Cole-Parmer Supplier No. 0612704
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.