Learn More
Abcam Anti-Strep-tag II antibody (GT661) Mouse monoclonal [GT661] to Strep-tag II; Suitable for: WB, IP, ICC/IF; Reacts with: Species independent; Isotype: IgG2a
Supplier:  Abcam AB184224100UL
Encompass_Preferred
Anti-Strep-tag II antibody (GT661) Mouse monoclonal [GT661] to Strep-tag II; Suitable for: WB, IP, ICC/IF; Reacts with: Species independent; Isotype: IgG2a
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2602128