Learn More
Seracare Life Sciences Inc ANTI-SWINE IGG (H+L) AB FITC
Supplier:  Seracare Life Sciences Inc 52300297
Encompass_Preferred
Affinity purified polyclonal antibody to Swine IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-668-83