Learn More
Rockland Immunochemicals Swine IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943940
Encompass_Preferred
Anti-Swine IgG whole molecule antibody generated in rabbit detects specifically Swine IgG whole molecule. This secondary antibody anti-Swine is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3940 Supplier Rockland Immunochemicals Supplier No. 6144302
Anti-Swine IgG whole molecule antibody generated in rabbit detects specifically Swine IgG whole molecule. This secondary antibody anti-Swine is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.