Learn More
Rockland Immunochemicals Swine IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Supplier:  Rockland Immunochemicals 6141302
Encompass
Anti-Swine IgG whole molecule antibody generated in goat detects specifically Swine IgG whole molecule. This secondary antibody anti-Rat is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3939