Learn More
Seracare Life Sciences Inc ANTI-SWINE IGM (MU) ANTIBODY
Catalog No. 5066814
Encompass_Preferred
Affinity purified polyclonal antibody to Swine IgM heavy chain (mu) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-668-14 Supplier Seracare Life Sciences Inc Supplier No. 52100172
Affinity purified polyclonal antibody to Swine IgM heavy chain (mu) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.