Learn More
ECM Bioscience Syntaphilin (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SM3711
Encompass_Preferred
Clone M371 was generated from a sequence corresponding to amino acids in the N-terminal region of human syntaphilin. This sequence has high homology to mouse and rat syntaphilin. Accession #O15079. This antibody detects a 70 kDa* protein corresponding to the molecular mass of Syntaphilin on SDS-PAGE immunoblots of adult mouse brain. Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal, protein A purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names = SNPH
Catalog No. 50-206-8852