Learn More
Immunology Consultants anti-T7 Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 5000435443
Encompass_Preferred
This affinity‑purified rabbit anti‑T7 antibody provides highly specific recognition of T7‑tagged recombinant proteins across a wide range of molecular biology applications. Antigen‑specific purification ensures strong binding and minimal background, making it ideal for Western blotting, ELISA, immunoprecipitation, and protein expression analysis. Manufactured by Immunology Consultants, this 1 mg preparation delivers consistent performance in both native and denatured samples. Its clean profile and robust affinity support reliable detection and quantification of T7‑tagged constructs, making it a dependable reagent for laboratories working with T7 epitope tagging systems.
Catalog No. 50-004-35443 Supplier Immunology Consultants Supplier No. RT745A
This affinity‑purified rabbit anti‑T7 antibody provides highly specific recognition of T7‑tagged recombinant proteins across a wide range of molecular biology applications. Antigen‑specific purification ensures strong binding and minimal background, making it ideal for Western blotting, ELISA, immunoprecipitation, and protein expression analysis. Manufactured by Immunology Consultants, this 1 mg preparation delivers consistent performance in both native and denatured samples. Its clean profile and robust affinity support reliable detection and quantification of T7‑tagged constructs, making it a dependable reagent for laboratories working with T7 epitope tagging systems.