Learn More
Immunology Consultants anti-T7 Antibody (Rabbit) - Affinity Purified
SDP
Supplier:  Immunology Consultants RT745AZ
Encompass_Preferred
This 0.1 mg format of the affinity‑purified anti‑T7 antibody offers the same high specificity and low background performance as the full‑size version in a compact, economical quantity. Ideal for pilot studies, assay optimization, or limited‑sample workflows, it provides reliable detection of T7‑tagged proteins in Western blotting, ELISA, and immunoprecipitation. Produced by Immunology Consultants, the antibody maintains strong epitope recognition and consistent performance across diverse assay formats. Its smaller size makes it a practical choice for researchers evaluating T7‑tag detection strategies without committing to a full milligram.
Catalog No. 50-004-35444