Learn More
Immunology Consultants anti-T7 Antibody (Rabbit) - HRP Conjugated
SDP
Catalog No. 5000435317
Encompass_Preferred
This HRP‑conjugated anti‑T7 antibody provides sensitive enzymatic detection of T7‑tagged proteins in Western blotting, ELISA, and chemiluminescent or colorimetric assays. The HRP label enables strong signal amplification, supporting detection of low‑abundance proteins and high‑contrast assay results. Manufactured by Immunology Consultants, this 1 mg reagent maintains excellent specificity for the T7 epitope and performs consistently across assay platforms. It is a reliable choice for researchers requiring robust, enzyme‑linked detection of T7‑tagged constructs in protein expression and characterization workflows
Catalog No. 50-004-35317 Supplier Immunology Consultants Supplier No. RT745P
This HRP‑conjugated anti‑T7 antibody provides sensitive enzymatic detection of T7‑tagged proteins in Western blotting, ELISA, and chemiluminescent or colorimetric assays. The HRP label enables strong signal amplification, supporting detection of low‑abundance proteins and high‑contrast assay results. Manufactured by Immunology Consultants, this 1 mg reagent maintains excellent specificity for the T7 epitope and performs consistently across assay platforms. It is a reliable choice for researchers requiring robust, enzyme‑linked detection of T7‑tagged constructs in protein expression and characterization workflows