Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cosmo Bio Usa Inc Anti TAR DNA-Binding Protein 43, phospho-Ser409/410 monoclonal antibody, Clone 11-9, 100 μL
SDP

Supplier:  Cosmo Bio Usa Inc CACTIPPTDM01A

Encompass

The COSMO BIO USA INC Anti TAR DNA-Binding Protein 43 (TDP-43) monoclonal antibody (Clone 11-9) is a highly specific reagent for detecting abnormally phosphorylated TDP-43 in human samples and serves as a vital tool for research on neurodegenerative diseases like Frontotemporal Lobar Degeneration and Amyotrophic Lateral Sclerosis. This antibody is designed for biochemical and immunohistochemical analyses, providing robust performance in various applications.

  • Produced in liquid PBS formulation
  • High specificity for phosphorylated TDP-43
  • Validated applications include Western Blot, ELISA, and Immunohistochemistry
  • Concentration range of 1.0-1.5 mg/mL
  • Volume of 100 μL
  • Storage recommended below -20°C
  • Research use only

Catalog No. NC2204971


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.