Learn More
Medchemexpress LLC Anti-TCR Antibody N 1mg | 1mg
SDP
Catalog No. 5000324750
Encompass_Preferred
Anti-TCR Antibody (NKTT320) is a humanized antibody expressed in CHO targeting TCR Anti-TCR Antibody (NKTT320) features huIgG4 heavy chain and hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for Anti-TCR Antibody (NKTT320) can refer to Human IgG4 kappa Isotype Control (HY-P99003)
Catalog No. 50-003-24750 Supplier Medchemexpress LLC Supplier No. HYP9905151MG
Anti-TCR Antibody (NKTT320) is a humanized antibody expressed in CHO targeting TCR Anti-TCR Antibody (NKTT320) features huIgG4 heavy chain and hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for Anti-TCR Antibody (NKTT320) can refer to Human IgG4 kappa Isotype Control (HY-P99003)