Learn More
ANCELL CORPORATION anti-TCR Cβ1, 120 tests
Supplier:  ANCELL CORPORATION 101040
Encompass
Human T cell receptor (TCR) is a T cell surface expressed heterodimer of alpha/beta or gamma/delta chains. TCR’s recognize antigen peptides bound to MHC molecules forming the basis of antigen specific response by T cells.
Catalog No. NC1648616