Learn More
ABclonal Technology TEAD1 Polyclonal Antibody,100UL,Validated in: WB, IHC, IF, ChIP
SDP
Supplier:  ABclonal Technology A13366
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 200-300 of human TEAD1 (NP_068780.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC,IF,ChIP•Swiss Prot: P28347•Gene ID:7003•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-164-8983