Learn More
Boca Scientific Inc TET03 Anti-Tet-Repressor Antibody, Monoclonal Mouse IgG1, lyophilized, 50µg
Supplier:  Boca Scientific Inc TET03
Encompass
Anti-Tet-Repressor (TetR) antibodies are a prime tool for studying tet regulatory systems in eukaryotic cells. TET03 is a mouse monoclonal antibody, IgG1 Kappa. The antibody recognizes amino acid # 37-44 and is ideal for immunofluorescence studies. TET03 is provided as 50µg of purified antibody, lyophilized. After reconstitution to 100µl in distilled water, it can be further diluted 1:100 to 1:500 for immunofluorescence studies.
Catalog No. NC0515501