Learn More
Immunology Consultants anti-Thyroxine (T4) Antibody (Mouse - Clone #100) - Monoclonal
SDP
Catalog No. 5000435400
Encompass_Preferred
This monoclonal antibody provides high‑specificity detection of thyroxine (T4), a major circulating thyroid hormone essential for metabolic and developmental regulation. Clone 100 offers strong affinity and minimal cross‑reactivity with T3 or other related molecules, supporting competitive immunoassays, ELISA, and hormone‑monitoring applications. MT4‑65A‑100 is ideal for thyroid research, endocrine diagnostics, and studies examining T4 regulation. Its reproducible performance ensures dependable detection across diverse sample types.
Catalog No. 50-004-35400 Supplier Immunology Consultants Supplier No. MT465A100
This monoclonal antibody provides high‑specificity detection of thyroxine (T4), a major circulating thyroid hormone essential for metabolic and developmental regulation. Clone 100 offers strong affinity and minimal cross‑reactivity with T3 or other related molecules, supporting competitive immunoassays, ELISA, and hormone‑monitoring applications. MT4‑65A‑100 is ideal for thyroid research, endocrine diagnostics, and studies examining T4 regulation. Its reproducible performance ensures dependable detection across diverse sample types.