Learn More
ABclonal Technology TMEM173 Polyclonal Antibody, 100UL, Validated in:WB, IHC, IP
SDP
Supplier:  ABclonal Technology A3575
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 150-379 of human STING/TMEM173 (NP_938023.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC,IF,IP•Swiss Prot: Q86WV6•Gene ID:340061•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-7555