Learn More
ABclonal Technology TPSAB1 Polyclonal Antibody, 100UL, Validated in:WB, IF
SDP
Supplier:  ABclonal Technology A2022
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TPSAB1 (NP_003285.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB•Swiss Prot: Q15661•Gene ID:7177•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-7018