Learn More
Genscript Corporation Anti-Trastuzumab Antibody (11C4), mAb, Mouse
Supplier:  Genscript Corporation A02032
Encompass_Preferred
The product is specific for Trastuzumab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A02033-40, Anti-Trastuzumab Antibody (15H2), mAb, Mouse.
Catalog No. NC2223968