Learn More
Sino Biological Anti-TrkA Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000785900
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TrkA (rR TrkA; Catalog#80404-R08H; NP_067600.1; Met1-Pro418). TrkA specific IgG was purified by Rat TrkA affinity chromatography.
Catalog No. 50-007-85900 Supplier Sino Biological Supplier No. 80404T16100
Produced in rabbits immunized with purified, recombinant Rat TrkA (rR TrkA; Catalog#80404-R08H; NP_067600.1; Met1-Pro418). TrkA specific IgG was purified by Rat TrkA affinity chromatography.