Learn More
STEMCELL Technologies Anti-Tyrosine Hydroxylase Antibody, Clone TH-2, Size: 200 µL
SDP
Supplier:  STEMCELL Technologies 60058
Encompass
Clone TH-2 is a mouse monoclonal antibody that reacts with human, rat, and cow tyrosine hydroxylase (TH). TH catalyzes the hydroxylation of L-tyrosine to L-3,4 dihydroxyphenylalanine (L-dopa) in the brain and adrenal medulla and can therefore be used to detect dopaminergic neurons. Verified for use with immunocytochemistry assays, use this antibody in applications requiring the detection of TH. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5315
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov