Learn More
ECM Bioscience unphosphorylated βIII-Tubulin (Ser-444) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience TP1811
Encompass_Preferred
This antibody was cross-adsorbed to phospho-βIII-Tubulin (Ser-444) peptide before affinity purification using unphosphorylated βIII-Tubulin (Ser-444) peptide (without carrier). The antibody detects a 50 kDa* protein corresponding to the molecular mass of unphosphorylated βIII-Tubulin on SDS-PAGE immunoblots of purified brain tubulin and mouse brain tissue. The immunogen was Unphosphorylated βIII-Tubulin (Ser-444) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 444 of human βIII-Tubulin. This sequence is not found in bI or βII-Tubulin isotypes, but is well conserved in βIII-Tubulins from rat and mouse. Accession #Q13509. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Tubulin bIII
Catalog No. 50-206-8866