Learn More
Sino Biological Anti-Urokinase/uPA Antibody (Biotin), Rabbit Polyclonal 100 µg
Catalog No. 5000749417
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Urokinase/uPA (rh Urokinase/uPA; Catalog#10815-H08H; NP_002649.1; Met1-Leu431). Urokinase/uPA specific IgG was purified by Human Urokinase/uPA affinity chromatography and then biotinylated.
Catalog No. 50-007-49417 Supplier Sino Biological Supplier No. 10815T20B100
Produced in rabbits immunized with purified, recombinant Human Urokinase/uPA (rh Urokinase/uPA; Catalog#10815-H08H; NP_002649.1; Met1-Leu431). Urokinase/uPA specific IgG was purified by Human Urokinase/uPA affinity chromatography and then biotinylated.