Learn More
ECM Bioscience VASP (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience VM2771
Encompass_Preferred
Clone (M277) was generated from a recombinant protein that includes amino acids from the C-terminal region of human VASP. Accession #P50552. This antibody detects 46 and 50 kDa* proteins corresponding to the apparent molecular mass of VASP on SDS-PAGE immunoblots of human A431, HeLa, and HUVEC. Applications: WB, E, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = vasodilator-stimulated phosphoprotein
Catalog No. 50-206-8870