Learn More
ECM Bioscience VE-Cadherin (a.a.770-781) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CP2231
Encompass_Preferred
This antibody was affinity purified with VE-cadherin (a.a. 770-781) peptide. The purified antibody detects a 140 kDa* band corresponding to VE-cadherin in western blots of human endothelial cells, and this reactivity can be specifically blocked using VE-cadherin peptide (CX2235). The immunogen was VE-Cadherin synthetic peptide (coupled to carrier protein) corresponds to amino acids 770 to 781 in human VE-cadherin. This sequence has significant homology to the conserved site in rat and mouse, and has less than 50% homology with other cadherins. Accession #P33151. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Cadherin-5, vascular endothelial Cadherin, CD144
Catalog No. 50-206-8680