Learn More
Aves Labs, Inc Anti-VGlut1 Antibody
Catalog No. NC2736791
Encompass
Anti-VGlut1 mouse monoclonal primary antibody from NeuroMab is produced in-house from hybridoma clone N28/9. It detects fish, human, mouse, and rat VGlut1, and is purified by Protein A chromatography. It is great for use in Flow, IHC, ICC, WB.
Catalog No. NC2736791 Supplier Aves Labs, Inc Supplier No. 7506602020UL
Anti-VGlut1 mouse monoclonal primary antibody from NeuroMab is produced in-house from hybridoma clone N28/9. It detects fish, human, mouse, and rat VGlut1, and is purified by Protein A chromatography. It is great for use in Flow, IHC, ICC, WB.