Learn More
FUJIFILM Biosciences Anti VGLUT1 Guinea Pig 50uL
Catalog No. 5000450131
Encompass_Preferred
Vesicular glutamate transporters VGLUT are involved in the transport of glutamate a major excitatory neurotransmitter into synaptic vesicles It is known that glutamate release associated with neural excitation contributes to various physiological phenomena such as cognitive function and memory VGLUT1 is primarily expressed in the cerebral cortex and hippocampus of the central nervous system and is used as a specific marker for glutamate-responsive neurons aiding in the identification of excitatory neurons This product is a guinea pig polyclonal antibody that recognizes VGLUT1 Its useful for glutamatergic neuron marker Specific to VGLUT1
Catalog No. 50-004-50131 Supplier FUJIFILM Biosciences Supplier No. 01328811
Vesicular glutamate transporters VGLUT are involved in the transport of glutamate a major excitatory neurotransmitter into synaptic vesicles It is known that glutamate release associated with neural excitation contributes to various physiological phenomena such as cognitive function and memory VGLUT1 is primarily expressed in the cerebral cortex and hippocampus of the central nervous system and is used as a specific marker for glutamate-responsive neurons aiding in the identification of excitatory neurons This product is a guinea pig polyclonal antibody that recognizes VGLUT1 Its useful for glutamatergic neuron marker Specific to VGLUT1