Learn More
Electron Microscopy Sciences AMCU High Load Anti-Vibration Platform with Stainless Steel Surface 75 x 90 cm
SDP
Catalog No. 501930691
Encompass_Preferred
High Performance anti-vibration platforms for instruments. The seamless design and stainless steel featured in the AMCU series makes these platforms ideal for use in laboratories and cleanrooms. Integrated cushion air isolators remove 95% of vibrations. These platforms are specifically designed for high load applications and will support weights up to 1000kgs. Typical applications include large spectrometers, high-load weighing machines, and material testing machines.
Size: 75 x 90 cm
Surface: Stainless Steel
Catalog No. 50-193-0691 Supplier Electron Microscopy Sciences Supplier No. 6954
High Performance anti-vibration platforms for instruments. The seamless design and stainless steel featured in the AMCU series makes these platforms ideal for use in laboratories and cleanrooms. Integrated cushion air isolators remove 95% of vibrations. These platforms are specifically designed for high load applications and will support weights up to 1000kgs. Typical applications include large spectrometers, high-load weighing machines, and material testing machines.
Size: 75 x 90 cm
Surface: Stainless Steel