Learn More
Electron Microscopy Sciences AMP Passive Air Isolation Platformwith Stainless Steel Surface 60 x 120 cm
SDP
Catalog No. 501930681
Encompass_Preferred
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 60 x 120 cm
Surface: Stainless Steel
Catalog No. 50-193-0681 Supplier Electron Microscopy Sciences Supplier No. 6350SS
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 60 x 120 cm
Surface: Stainless Steel