Learn More
Grainger Anti-Vibration Gloves 2XL Black/Ylw PR
Catalog No. 502764138
Encompass_Preferred
The Grainger Approved Anti-Vibration Gloves are designed to reduce vibration transmitted to the hands and fingers while providing abrasion resistance. They feature a soft, breathable Lycra spandex back with hi-viz color panels and an ergonomic neoprene palm with gel padding to absorb shock and vibration. The gloves have an elastic wrist to keep dirt and debris out. They come in sizes S-2XL and are ideal for applications like construction, landscaping, manufacturing, and automotive repair where vibration damping is needed. The gloves meet ANSI/ISEA 105-2016 standards.
Catalog No. 50-276-4138 Supplier Grainger Supplier No. 16P217
The Grainger Approved Anti-Vibration Gloves are designed to reduce vibration transmitted to the hands and fingers while providing abrasion resistance. They feature a soft, breathable Lycra spandex back with hi-viz color panels and an ergonomic neoprene palm with gel padding to absorb shock and vibration. The gloves have an elastic wrist to keep dirt and debris out. They come in sizes S-2XL and are ideal for applications like construction, landscaping, manufacturing, and automotive repair where vibration damping is needed. The gloves meet ANSI/ISEA 105-2016 standards.