Learn More
Adam Equipment Anti-vibration table
Catalog No. NC1091130
Encompass_Preferred
Adam Equipments AVT is Ergonomically styled. The AVT anti vibration table offers a solid, stable surface developed specifically to help decrease vibration during measurement. When used in tandem with a weighing balance, the table allows the balance to perform with marked precision, despite air currents or movements that might cause readings to fluctuate. The AVT is easy to assemble, with all necessary tools supplied. The internal working surface measures 15.7” x 17.7” / 400 x 450mm, allowing ample space for a balance. Its large, scratch-resistant dark granite slab sits on shock-absorbing rubber mounts, while levelling feet accommodate uneven surfaces. The balance fits in the middle of the AVT’s granite surface, stabilizing the base and enabling greater precision.Recommended for use alongside our Equinox, Solis and Luna ranges of analytical balances.
Catalog No. NC1091130 Supplier Adam Equipment Supplier No. 104008036
Adam Equipments AVT is Ergonomically styled. The AVT anti vibration table offers a solid, stable surface developed specifically to help decrease vibration during measurement. When used in tandem with a weighing balance, the table allows the balance to perform with marked precision, despite air currents or movements that might cause readings to fluctuate. The AVT is easy to assemble, with all necessary tools supplied. The internal working surface measures 15.7” x 17.7” / 400 x 450mm, allowing ample space for a balance. Its large, scratch-resistant dark granite slab sits on shock-absorbing rubber mounts, while levelling feet accommodate uneven surfaces. The balance fits in the middle of the AVT’s granite surface, stabilizing the base and enabling greater precision.Recommended for use alongside our Equinox, Solis and Luna ranges of analytical balances.