Learn More
Braintree Scientific Anti-Vibration Table, 15.7" x 17.7" / 400 x 450mm
SDP
Catalog No. 501954167
Encompass
Anti-Vibration Table - Ergonomically styled, the anti-vibration table offers a solid, stable surface developed specifically to help decrease vibration during measurement. When used in tandem with a weighing balance, the table allows the balance to perform with marked precision, despite air currents or movements that might cause readings to fluctuate. Easy to assemble, with all necessary tools supplied. The internal working surface measures 15.7 x 17.7 / 400 x 450mm, allowing ample space for a balance. Its large, scratch-resistant dark granite slab sits on shock-absorbing rubber mounts, while levelling feet accommodate uneven surfaces. The balance fits in the middle of the granite surface, stabilizing the base and enabling greater precision. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4167 Supplier Braintree Scientific Supplier No. ADVT
Anti-Vibration Table - Ergonomically styled, the anti-vibration table offers a solid, stable surface developed specifically to help decrease vibration during measurement. When used in tandem with a weighing balance, the table allows the balance to perform with marked precision, despite air currents or movements that might cause readings to fluctuate. Easy to assemble, with all necessary tools supplied. The internal working surface measures 15.7 x 17.7 / 400 x 450mm, allowing ample space for a balance. Its large, scratch-resistant dark granite slab sits on shock-absorbing rubber mounts, while levelling feet accommodate uneven surfaces. The balance fits in the middle of the granite surface, stabilizing the base and enabling greater precision. **Sold for research purposes only and not for human use**