Learn More
Medchemexpress LLC HY-131233 5mg Medchemexpress, Anti-virus agent 1 CAS:1911578-83-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1312335MG
Encompass_Preferred
Medchemexpress, HY-131233 5mg Anti-virus agent 1 CAS:1911578-83-0 Anti-virus agent 1 (compound 4i), a phosphoramidate prodrug of GS-5734 (HY-104077; Remdesivir), has potent antiviral activity. Anti-virus agent 1 is used for the research of coronavirus and Ebola virus (EBOV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1269