Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-Virus Compound Library, 250uL/well(10 mM solution),96 Well Racks With Screw Cap
SDP
Supplier:  Apexbio Technology LLC L1050.25
Encompass_Preferred
DiscoveryProbe™ Anti-Virus Compound Library is a collection of 264 Anti-Virus compounds supplied as lyophilized powder or pre-dissolved DMSO solutions. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8490
Add to Cart
Request a Quote