Learn More
ECM Bioscience WAVE1 (N-terminal region) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience WP1731
Encompass_Preferred
This antibody was affinity purified using WAVE1 (N-terminal region) peptide (without carrier). The antibody detects an 80 kDa* protein corresponding to the molecular mass of WAVE1 on SDS-PAGE immunoblots of human SYF cSrc-transformed, HUVEC, and K562 cells, as well as rat PC12 cells and mouse C2C12 cells. The immunogen was A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal half of human WAVE1. This sequence has high homology with similar regions in rat and mouse WAVE1, and has less than 50% homology to similar regions in human WAVE2 and WAVE3. Accession #Q92558. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Wiskott-Aldrich syndrome verproline, Scar1, WASF1
Catalog No. 50-206-8876