Learn More
BIOHIPPO Anti-West Nile virus/WNV GP1/Genome polyprotein Antibody (CR4274), 100 µg
SDP
Catalog No. 5000860930
Encompass_Preferred
West Nile virus/WNV GP1/Genome polyprotein Recombinant Antibody (clone CR4274) is a recombinant Human IgG1, kappa antibody specific for West Nile virus/WNV GP1/Genome polyprotein. It reacts with West Nile virus. Suitable for ELISA, SPR, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60930 Supplier BIOHIPPO Supplier No. BHA21405497
West Nile virus/WNV GP1/Genome polyprotein Recombinant Antibody (clone CR4274) is a recombinant Human IgG1, kappa antibody specific for West Nile virus/WNV GP1/Genome polyprotein. It reacts with West Nile virus. Suitable for ELISA, SPR, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.