Learn More
Biolegend Purified anti-mouse/rat XCR1
Supplier:  Biolegend 148202
Encompass_Preferred
Purified anti-mouse/rat XCR1 [ZET]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 100 ug
Catalog No. NC1365855