Learn More
Sino Biological Anti-XK Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000704808
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human XK fragment, and purified by antigen affinity chromatography.
Catalog No. 50-007-04808 Supplier Sino Biological Supplier No. 204540T02100
Produced in rabbits immunized with E. coli-derived Human XK fragment, and purified by antigen affinity chromatography.