Learn More
BIOHIPPO Anti-Zika Virus Envelope protein E Nanobody (SAA2297), 100 µg
SDP
Catalog No. 5000859732
Encompass_Preferred
Zika Virus Envelope E Nanobody Recombinant Antibody (clone SAA2297) is a recombinant Alpaca VHH-His antibody specific for Zika Virus Envelope E Nanobody. It reacts with Zika Virus. Suitable for ELISA, WB, FCM. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-59732 Supplier BIOHIPPO Supplier No. BHA21405020
Zika Virus Envelope E Nanobody Recombinant Antibody (clone SAA2297) is a recombinant Alpaca VHH-His antibody specific for Zika Virus Envelope E Nanobody. It reacts with Zika Virus. Suitable for ELISA, WB, FCM. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.