Learn More
Biomatik Corporation 5-Hydroxytryptamine (5-HT) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11929
Encompass_Preferred
5-Hydroxytryptamine (5-HT) Anti-antigen ELISA Kit for detecting 5-HT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: 5HT; Serotonin; N-Methyl-Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8064